Gene Transcripts

Gene Symbol
Slc35g1
Entrez Gene ID
240660
Gene Name
solute carrier family 35, member G1
Gene Aliases
AA590464, D330039I19Rik, Tmem20
Chromosome Location
Chr. 19: 38395980 - 38405607 on Build GRCm38
UniGene ID
Mm.262085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175507.3 NP_780716.1 3 1083
GenBank mRNA AK041618.1 - 3 1083
AK052365.1 - 3 987
AK171987.1 - 3 1015
BC137663.1 - 3 1094 8860682
BC137664.1 - 3 1094 8860683

View Full Product Details