Gene Transcripts

Gene Symbol
Exph5
Entrez Gene ID
320051
Gene Name
exophilin 5
Gene Aliases
AC079869.22gm5, B130009M24Rik, E030050P12, Kiaa0624, Slac2b, slac2-b
Chromosome Location
Chr. 9: 53301519 - 53381166 on Build GRCm38
UniGene ID
Mm.277540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176846.3 NP_789816.2 3 424
GenBank mRNA AB098163.1 - 3 424
BC120905.1 - 3 411 40087301

View Full Product Details