Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr. 12: 49322135 - 49331733 on Build GRCh38
UniGene ID
Hs.524399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005480.3 NP_005471.3 12 1369
XM_006719181.1 XP_006719244.1 12 1313
XM_011537724.1 XP_011536026.1 11 1109
GenBank mRNA AK315211.1 - 12 1313
BC032583.1 - 12 1281 5498800
U04810.2 - 12 1312

View Full Product Details