Gene Transcripts

Gene Symbol
Rgl3
Entrez Gene ID
71746
Gene Name
ral guanine nucleotide dissociation stimulator-like 3
Gene Aliases
1300003D20Rik, mFLJ00153
Chromosome Location
Chr. 9: 21971179 - 21989465 on Build GRCm38
UniGene ID
Mm.110594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023622.4 NP_076111.2 5 568
GenBank mRNA AF237669.1 - 5 527
AF239661.1 - 5 569
AK004876.1 - 4 301
AK053550.1 - 5 555
AK134654.1 - 550
AK165023.1 - 5 556
AK165746.1 - 5 561

View Full Product Details