Gene Transcripts
- Gene Symbol
- RGS4
- Entrez Gene ID
- 5999
- Gene Name
- regulator of G-protein signaling 4
- Gene Aliases
- RGP4, SCZD9
- Chromosome Location
- Chr. 1: 163068606 - 163076802 on Build GRCh38
- UniGene ID
- Hs.386726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102445.2 |
NP_001095915.1
|
5, 6 |
994 |
|
NM_001113380.1 |
NP_001106851.1
|
4, 5 |
569 |
|
NM_005613.5 |
NP_005604.1
|
4, 5 |
885 |
|
GenBank mRNA |
AF493928.1 |
- |
4, 5 |
374 |
|
AK295540.1 |
- |
5, 6 |
729 |
|
AK296240.1 |
- |
4, 5 |
413 |
|
AK300832.1 |
- |
4, 5 |
373 |
|
AK312250.1 |
- |
4, 5 |
497 |
|
BC000737.1 |
- |
4, 5 |
459 |
3510260 |
BC051869.1 |
- |
4, 5 |
471 |
5786695 |
BI547160.1 |
- |
4, 5 |
599 |
|
BT007025.1 |
- |
4, 5 |
374 |
|
DQ346662.1 |
- |
4, 5 |
498 |
|
DQ346663.1 |
- |
4, 5 |
713 |
|
DQ346664.1 |
- |
5, 6 |
825 |
|
EF054877.1 |
- |
4, 5 |
569 |
|
U27768.1 |
- |
4, 5 |
471 |
|
View Full Product Details