Gene Transcripts

Gene Symbol
Mettl14
Entrez Gene ID
210529
Gene Name
methyltransferase like 14
Gene Aliases
G430022H21Rik, mKIAA1627
Chromosome Location
Chr. 3: 123368295 - 123385990 on Build GRCm38
UniGene ID
Mm.260039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201638.2 NP_964000.2 10 1126
GenBank mRNA AK146881.1 - 10 1115
AK153810.1 - 10 1122
BC044899.1 - 1 1883
BC052204.1 - 10 1084 30007932

View Full Product Details