Gene Transcripts

Gene Symbol
Fchsd1
Entrez Gene ID
319262
Gene Name
FCH and double SH3 domains 1
Gene Aliases
A030002D08Rik
Chromosome Location
Chr. 18: 37957434 - 37969814 on Build GRCm38
UniGene ID
Mm.285060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175684.4 NP_783615.2 7 526
GenBank mRNA AK154732.1 - 7 560
BC052886.1 - 7 557
BC057367.1 - 7 526 6827858

View Full Product Details