Gene Transcripts

Gene Symbol
Spen
Entrez Gene ID
56381
Gene Name
SPEN homolog, transcriptional regulator (Drosophila)
Gene Aliases
Mint, mKIAA0929
Chromosome Location
Chr. 4: 141467885 - 141538635 on Build GRCm38
UniGene ID
Mm.299906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019763.2 NP_062737.2 2 635
GenBank mRNA AB055980.1 - 1 80
AF156529.1 - 1 140
AK137647.1 - 2 523

View Full Product Details