Gene Transcripts

Gene Symbol
Skint8
Entrez Gene ID
639774
Gene Name
selection and upkeep of intraepithelial T cells 8
Gene Aliases
OTTMUSG00000009475
Chromosome Location
Chr. 4: 111919446 - 111950361 on Build GRCm38
UniGene ID
Mm.248185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100466.1 NP_001093936.2 3 358
GenBank mRNA AK076260.1 - 2 2413
EF494903.1 - 3 358
EF494904.1 - 3 358
EU099307.1 - 3 358
EU099308.1 - 3 358

Gene Symbol
Skint7
Entrez Gene ID
328505
Gene Name
selection and upkeep of intraepithelial T cells 7
Gene Aliases
C130057D23Rik
Chromosome Location
Chr. 4: 111972924 - 111988260 on Build GRCm38
UniGene ID
Mm.226771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142775.1 NP_001136247.1 3 510
NM_177818.3 NP_808486.2 3 510
GenBank mRNA AK048392.1 - 3 405
AK132639.1 - 3 515
BB614300.1 - 3 510
EF494899.1 - 348
EF494900.1 - 348
EF494901.1 - 348
EF494902.1 - 348
EU099306.1 - 348

View Full Product Details