Gene Transcripts

Gene Symbol
Fsd1
Entrez Gene ID
240121
Gene Name
fibronectin type 3 and SPRY domain-containing protein
Gene Aliases
Chromosome Location
Chr. 17: 55986462 - 55996883 on Build GRCm38
UniGene ID
Mm.270660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183178.2 NP_899001.1 6 552
GenBank mRNA AK140132.1 - 6 586
AK144257.1 - 6 487
BC055106.1 - 6 552 6532001

View Full Product Details