Gene Transcripts

Gene Symbol
Ahdc1
Entrez Gene ID
230793
Gene Name
AT hook, DNA binding motif, containing 1
Gene Aliases
D030015G18Rik
Chromosome Location
Chr. 4: 133011265 - 133078110 on Build GRCm38
UniGene ID
Mm.31816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146155.3 NP_666267.3 6 2737
GenBank mRNA AK134372.1 - 1 290
BC042621.1 - 6 2726 4911638
BC060231.1 - 5 2705 6852595

View Full Product Details