Gene Transcripts

Gene Symbol
Poglut1
Entrez Gene ID
224143
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
9630046K23Rik, Clp46, Ktelc1, Rumi
Chromosome Location
Chr. 16: 38525058 - 38550256 on Build GRCm38
UniGene ID
Mm.284366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300827.1 NP_001287756.1 2 442
NM_172380.4 NP_759012.1 2 239
GenBank mRNA AK031608.1 - 2 178
AK035948.1 - 2 187
AK036224.1 - 2 169
AK045278.1 - 2 387
AK079886.1 - 2 208
AK082556.1 - 2 409
AK167641.1 - 2 409
AK197458.1 - 1 279
AK215456.1 - 2 81
AK216766.1 - 2 81
BC026809.1 - 2 190 4205469
CB249259.1 - 2 240

View Full Product Details