Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004196.3 | NP_001004196.2 | 4 | 460 | |
NM_001318826.1 | NP_001305755.1 | 3 | 385 | ||
NM_001318828.1 | NP_001305757.1 | 2 | 303 | ||
NM_001318830.1 | NP_001305759.1 | 2 | 303 | ||
NM_005944.6 | NP_005935.4 | 3 | 385 | ||
XM_005247481.4 | XP_005247538.1 | 4 | 634 | ||
XM_005247482.2 | XP_005247539.1 | 3 | 264 | ||
XM_005247485.2 | XP_005247542.1 | 3 | 575 | ||
XM_005247486.2 | XP_005247543.1 | 2 | 72 | ||
XM_005247488.2 | XP_005247545.1 | 2 | 182 | ||
XM_017006452.1 | XP_016861941.1 | 4 | 961 | ||
XM_017006453.1 | XP_016861942.1 | 2 | 227 | ||
XM_017006454.1 | XP_016861943.1 | 4 | 950 | ||
XM_017006455.1 | XP_016861944.1 | 3 | 585 | ||
XM_017006456.1 | XP_016861945.1 | 4 | 311 | ||
XM_017006457.1 | XP_016861946.1 | 3 | 264 | ||
XM_017006458.1 | XP_016861947.1 | 4 | 956 | ||
XM_017006459.1 | XP_016861948.1 | 3 | 575 | ||
GenBank mRNA | AF063591.1 | - | 3 | 176 | |
AK074972.1 | - | 3 | 150 | ||
AK129797.1 | - | 2 | 72 | ||
AK293399.1 | - | 2 | 72 | ||
AK297194.1 | - | 4 | 251 | ||
AK312563.1 | - | 3 | 266 | ||
AL832529.1 | - | 3 | 170 | ||
AY603771.1 | - | 2 | 119 | ||
BC022522.1 | - | 3 | 178 | 4794050 | |
BC031103.1 | - | 3 | 154 | 5301702 | |
BI547787.1 | - | 4 | 256 | ||
DA544134.1 | - | 3 | 263 |