Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178697.4 | NP_848812.1 | 5 | 758 | |
GenBank mRNA | AK028704.1 | - | 5 | 759 | |
AK035512.1 | - | 5 | 825 | ||
AK048276.1 | - | 5 | 749 | ||
AY161007.1 | - | 5 | 714 | ||
BC096379.1 | - | 5 | 821 | 5317084 |