Gene Transcripts

Gene Symbol
HS3ST3A1
Entrez Gene ID
9955
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Gene Aliases
3-OST-3A, 3OST3A1
Chromosome Location
Chr. 17: 13494032 - 13601942 on Build GRCh38
UniGene ID
Hs.462270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006042.2 NP_006033.1 2 2348
XM_011524114.2 XP_011522416.1 3 2592
GenBank mRNA AF105376.1 - 2 2333

View Full Product Details