Gene Transcripts

Gene Symbol
Olfr78
Entrez Gene ID
170639
Gene Name
olfactory receptor 78
Gene Aliases
4633402A21Rik, MOL2.3, MOR18-2, Or51e2, PSGR, RA1c
Chromosome Location
Chr. 7: 102740721 - 102759471 on Build GRCm38
UniGene ID
Mm.184372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168503.1 NP_001161975.1 3 1054
NM_130866.4 NP_570936.1 4 1923
GenBank mRNA AF378854.1 - 1 708
AK028467.1 - 4 1848
AK036356.1 - 3 1055
BC120603.1 - 2 897 40129526
BC120629.1 - 2 897 40129552

View Full Product Details