Gene Transcripts

Gene Symbol
Vmn1r67
Entrez Gene ID
171263
Gene Name
vomeronasal 1 receptor 67
Gene Aliases
V1re10
Chromosome Location
Chr. 7: 10446789 - 10447787 on Build GRCm38
UniGene ID
Mm.377216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134229.2 NP_598990.2 1 513
GenBank mRNA BC138174.1 - 1 513
BC145742.1 - 1 513 40131045

View Full Product Details