Gene Transcripts

Gene Symbol
Trp53bp2
Entrez Gene ID
209456
Gene Name
transformation related protein 53 binding protein 2
Gene Aliases
53BP2, AI746547, ASPP2, PPP1R13A, Tp53bp2, X98550
Chromosome Location
Chr. 1: 182409167 - 182462436 on Build GRCm38
UniGene ID
Mm.287450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173378.2 NP_775554.2 6, 7 900
GenBank mRNA AK189404.1 - 1, 2 133
BC042874.1 - 5, 6 502
CN529251.1 - 2, 3 188

View Full Product Details