Gene Transcripts

Gene Symbol
Rnf20
Entrez Gene ID
109331
Gene Name
ring finger protein 20
Gene Aliases
4833430L21Rik, AW540162, C79397, mKIAA4116
Chromosome Location
Chr. 4: 49632011 - 49656886 on Build GRCm38
UniGene ID
Mm.24765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163263.1 NP_001156735.1 4 909
NM_182999.2 NP_892044.1 5 756
GenBank mRNA AK048862.1 - 4 868
AK051278.1 - 4 868
AK053783.1 - 5 744
AK082529.1 - 4 813
AK085436.1 - 4 887
AK087860.1 - 4 835
AK163905.1 - 1 122
AK220492.1 - 4 825
BC052482.1 - 5 695 6308280

View Full Product Details