Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
79043
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
Chromosome Location
Chr. 17: 24886349 - 24892147 on Build GRCm38
UniGene ID
Mm.274794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163750.1 NP_001157222.1 7 1243
NM_001163751.1 NP_001157223.1 7 898
NM_027141.2 NP_081417.2 7 919
GenBank mRNA AF403038.1 - 7 919
AK009325.1 - 7 919
AK076170.1 - 7 1250
AK082768.1 - 7 842
AK156579.1 - 6 1256
AK167474.1 - 6 1236
AK220234.1 - 7 1181
BC026523.1 - 7 1584
BC072556.1 - 7 1204 5691114

View Full Product Details