Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028889.2 | NP_083165.1 | 4 | 1289 | |
GenBank mRNA | AK016491.1 | - | 4 | 1289 | |
AK045911.1 | - | 5 | 1166 | ||
AK144175.1 | - | 4 | 1292 | ||
AK171846.1 | - | 4 | 1291 | ||
BC019531.1 | - | 4 | 1275 | 4459208 | |
BC085088.1 | - | 4 | 1274 | 6467625 |