Gene Transcripts
- Gene Symbol
- Lrba
- Entrez Gene ID
- 80877
- Gene Name
- LPS-responsive beige-like anchor
- Gene Aliases
- C80285, D3Ertd775e, Lab300, Lba
- Chromosome Location
- Chr. 3: 86223568 - 86782705 on Build GRCm38
- UniGene ID
- Mm.439825
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF187731.1 |
- |
18 |
2539 |
|
AF188506.1 |
- |
18 |
2539 |
|
AF188507.1 |
- |
18 |
2539 |
|
View Full Product Details