Gene Transcripts

Gene Symbol
Tex11
Entrez Gene ID
83558
Gene Name
testis expressed gene 11
Gene Aliases
4930565P14Rik, BB024231, ZIP4H
Chromosome Location
Chr. X: 100838648 - 101059639 on Build GRCm38
UniGene ID
Mm.134024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167997.1 NP_001161469.1 9 813
NM_031384.2 NP_113561.2 9 813
GenBank mRNA AF285572.1 - 9 811
AK029799.1 - 9 814
AK082794.1 - 9 805
BC116709.1 - 9 809 40096708

View Full Product Details