Gene Transcripts

Gene Symbol
Pcdhgb2
Entrez Gene ID
93700
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
Chromosome Location
Chr. 18: 37689859 - 37841872 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033575.3 NP_291053.1 1 458
GenBank mRNA AY013805.1 - 1 359
BC054557.1 - 1 458 5706538

View Full Product Details