Gene Transcripts

Gene Symbol
Spag11a
Entrez Gene ID
78128
Gene Name
sperm associated antigen 11A
Gene Aliases
9230111C08Rik, Bin1b, EP2Q, EP2e, Spag11
Chromosome Location
Chr. 8: 19157214 - 19159920 on Build GRCm38
UniGene ID
Mm.229357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153115.1 NP_694755.1 2 371
GenBank mRNA AB089183.1 - 2 371
AF384173.1 - 2 334
AK020333.1 - 2 375

View Full Product Details