Gene Transcripts

Gene Symbol
Usp6nl
Entrez Gene ID
98910
Gene Name
USP6 N-terminal like
Gene Aliases
AI316785, RNTRE, TRE2NL, mKIAA0019
Chromosome Location
Chr. 2: 6322757 - 6444363 on Build GRCm38
UniGene ID
Mm.40673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080548.1 NP_001074017.1 7 580
NM_181399.3 NP_852064.2 6 419
GenBank mRNA AK084045.1 - 7 403
AK129037.1 - 7 432
AK155073.1 - 7 581
BC051184.1 - 6 422
BQ938063.1 - 6 419

View Full Product Details