Gene Transcripts

Gene Symbol
Kctd6
Entrez Gene ID
71393
Gene Name
potassium channel tetramerisation domain containing 6
Gene Aliases
5430433B02Rik, AU044285
Chromosome Location
Chr. 14: 8214148 - 8223570 on Build GRCm38
UniGene ID
Mm.292712
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017384.1 - 3 691
AK082924.1 - 3 690
AK159067.1 - 3 2103
BC096370.1 - 3 530 3597940
BC131971.1 - 2 500 40130248
BC131975.1 - 2 500 40130252

View Full Product Details