Gene Transcripts

Gene Symbol
Ubiad1
Entrez Gene ID
71707
Gene Name
UbiA prenyltransferase domain containing 1
Gene Aliases
1200002M06Rik, AI426463, AW320947, Tere1
Chromosome Location
Chr. 4: 148434497 - 148444751 on Build GRCm38
UniGene ID
Mm.292503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027873.2 NP_082149.1 2 1693
GenBank mRNA AK004550.1 - 2 1693
AK078442.2 - 2 1694
BC015303.1 - 2 1401 4241204
BC071203.1 - 2 1683 3667534

View Full Product Details