Gene Transcripts

Gene Symbol
Ttc4
Entrez Gene ID
72354
Gene Name
tetratricopeptide repeat domain 4
Gene Aliases
2810002P21Rik, AF007005, L62
Chromosome Location
Chr. 4: 106655014 - 106678967 on Build GRCm38
UniGene ID
Mm.25806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172073.1 NP_001165544.1 3 463
NM_028209.2 NP_082485.1 3 462
GenBank mRNA AK005923.1 - 3 464
AK012652.1 - 3 408
AK088684.1 - 3 424
AK133931.1 - 3 427
BC025435.1 - 3 411 5004463

View Full Product Details