Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006332.4 | NP_006323.2 | 3 | 436 | |
GenBank mRNA | AB049659.1 | - | 3 | 424 | |
AF097362.1 | - | 3 | 407 | ||
AK123477.1 | - | 2 | 1113 | ||
AK291551.1 | - | 3 | 421 | ||
AL558891.3 | - | 3 | 436 | ||
BC021136.1 | - | 3 | 405 | 4863531 | |
BC031020.1 | - | 3 | 403 | 4690088 | |
BE515053.1 | - | 3 | 553 |