Gene Transcripts

Gene Symbol
Iqcd
Entrez Gene ID
75732
Gene Name
IQ motif containing D
Gene Aliases
4933433C09Rik, AI662646
Chromosome Location
Chr. 5: 120586060 - 120607243 on Build GRCm38
UniGene ID
Mm.425285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029408.2 NP_083684.1 5 1310
GenBank mRNA AK017032.1 - 5 1310
BC125429.1 - 5 1304 40129944
BC137644.1 - 5 1304 8860663

View Full Product Details