Gene Transcripts
- Gene Symbol
- TUBGCP3
- Entrez Gene ID
- 10426
- Gene Name
- tubulin gamma complex associated protein 3
- Gene Aliases
- 104p, GCP3, Grip104, SPBC98, Spc98, Spc98p
- Chromosome Location
- Chr. 13: 112485005 - 112606469 on Build GRCh38
- UniGene ID
- Hs.224152
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286277.1 |
NP_001273206.1
|
7, 8 |
1018 |
|
NM_001286278.1 |
NP_001273207.1
|
7, 8 |
1048 |
|
NM_001286279.1 |
NP_001273208.1
|
7, 8 |
1048 |
|
NM_006322.5 |
NP_006313.1
|
7, 8 |
1048 |
|
XM_005268293.1 |
XP_005268350.1
|
7, 8 |
1043 |
|
XM_011537462.2 |
XP_011535764.1
|
7, 8 |
1001 |
|
XM_011537463.2 |
XP_011535765.1
|
8, 9 |
977 |
|
XM_011537464.2 |
XP_011535766.1
|
7, 8 |
1781 |
|
XM_017020323.1 |
XP_016875812.1
|
8, 9 |
1395 |
|
GenBank mRNA |
AF042378.1 |
- |
7, 8 |
927 |
|
AJ003061.1 |
- |
7, 8 |
843 |
|
AJ003062.1 |
- |
7, 8 |
843 |
|
AK302537.1 |
- |
7, 8 |
965 |
|
AK314785.1 |
- |
7, 8 |
1048 |
|
BC007763.2 |
- |
7, 8 |
998 |
|
BC013781.1 |
- |
7, 8 |
988 |
3867019 |
BC046634.2 |
- |
7, 8 |
1031 |
5295831 |
BT009772.1 |
- |
7, 8 |
843 |
|
View Full Product Details