Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172413.2 | NP_766001.1 | 3 | 2113 | |
GenBank mRNA | AK008416.1 | - | 1 | 1160 | |
AK087971.1 | - | 3 | 2060 | ||
AK146057.1 | - | 3 | 2093 | ||
AK146956.1 | - | 3 | 2155 | ||
AK147015.1 | - | 3 | 2155 | ||
BC027363.1 | - | 3 | 2077 | 4951566 | |
BC050056.1 | - | 3 | 2075 | 30016070 | |
BC064814.1 | - | 3 | 2109 | 6513822 | |
BC094890.1 | - | 3 | 2113 | 30649398 |