Gene Transcripts

Gene Symbol
Aasdhppt
Entrez Gene ID
67618
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 9: 4294793 - 4309509 on Build GRCm38
UniGene ID
Mm.33970
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026276.3 NP_080552.3 4 713
GenBank mRNA AK007737.1 - 4 714
AK008554.1 - 3 516
AK010557.1 - 4 673
AK013111.1 - 4 667
AK014188.1 - 4 656
AK018191.1 - 3 359
AK029523.1 - 4 655
AK031766.1 - 3 535
AK076099.1 - 4 668
AK076866.1 - 4 687
AK081818.1 - 4 690
AK088977.1 - 3 531
AK144671.1 - 4 687
BC030043.1 - 4 648 1348651
BC038013.1 - 4 684 3497754
BC049851.1 - 4 674 3481972

View Full Product Details