Gene Transcripts
- Gene Symbol
- March5
- Entrez Gene ID
- 69104
- Gene Name
- membrane-associated ring finger (C3HC4) 5
- Gene Aliases
- 1810015H18Rik, 2310008I22Rik, 2700055A20Rik, 5730499H23Rik, E130202O05Rik, MARCH-V, MITOL, Marchv, Rnf153
- Chromosome Location
- Chr. 19: 37207545 - 37224457 on Build GRCm38
- UniGene ID
- Mm.175989
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164336.1 |
NP_001157808.1
|
3, 4 |
702 |
|
NM_027314.3 |
NP_081590.3
|
3, 4 |
702 |
|
GenBank mRNA |
AK007511.1 |
- |
3, 4 |
538 |
|
AK009232.1 |
- |
3, 4 |
688 |
|
AK009364.1 |
- |
3, 4 |
700 |
|
AK178252.1 |
- |
2, 3 |
208 |
|
AK178944.1 |
- |
2, 3 |
216 |
|
AK206577.1 |
- |
1, 2 |
97 |
|
AK219813.1 |
- |
2, 3 |
164 |
|
BC064752.1 |
- |
3, 4 |
426 |
30049608 |
BC107215.1 |
- |
3, 4 |
414 |
40051628 |
BC132455.1 |
- |
3, 4 |
466 |
40130732 |
BC132457.1 |
- |
3, 4 |
465 |
40130734 |
View Full Product Details