Gene Transcripts

Gene Symbol
March5
Entrez Gene ID
69104
Gene Name
membrane-associated ring finger (C3HC4) 5
Gene Aliases
1810015H18Rik, 2310008I22Rik, 2700055A20Rik, 5730499H23Rik, E130202O05Rik, MARCH-V, MITOL, Marchv, Rnf153
Chromosome Location
Chr. 19: 37207545 - 37224457 on Build GRCm38
UniGene ID
Mm.175989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027314.3 NP_081590.3 6 1081
GenBank mRNA AK003478.1 - 2 126
AK009364.1 - 6 1079
AK188043.1 - 3 382
AK205563.1 - 2 148
BC064752.1 - 6 805 30049608
BC107214.1 - 6 788
BC107215.1 - 6 793 40051628
BC132455.1 - 6 845 40130732
BC132457.1 - 6 844 40130734

View Full Product Details