Gene Transcripts
- Gene Symbol
- Tra2a
- Entrez Gene ID
- 101214
- Gene Name
- transformer 2 alpha homolog (Drosophila)
- Gene Aliases
- 1500010G04Rik, AL022798, G430041M01Rik, mAWMS1
- Chromosome Location
- Chr. 6: 49243921 - 49264248 on Build GRCm38
- UniGene ID
- Mm.196598
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076230.1 |
- |
1 |
1484 |
|
AK076606.1 |
- |
1 |
1466 |
|
View Full Product Details