Gene Transcripts

Gene Symbol
Kif18b
Entrez Gene ID
70218
Gene Name
kinesin family member 18B
Gene Aliases
3000004C01Rik
Chromosome Location
Chr. 11: 102905518 - 102925134 on Build GRCm38
UniGene ID
Mm.32084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197959.2 NP_932063.2 2 266
GenBank mRNA AK156592.1 - 2 236
BC057614.1 - 2 234 6401536
BQ571943.1 - 2 229
BY312717.1 - 2 267

View Full Product Details