Gene Transcripts

Gene Symbol
DYNLT1
Entrez Gene ID
6993
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
CW-1, TCTEL1, tctex-1
Chromosome Location
Chr. 6: 158636474 - 158644786 on Build GRCh38
UniGene ID
Hs.445999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291602.1 NP_001278531.1 4 625
NM_001291603.1 NP_001278532.1 5 647
NM_006519.3 NP_006510.1 5 667
GenBank mRNA AK026669.1 - 2 2791
BC029412.1 - 5 620 4074224
BC105588.1 - 5 616 6723966
D50663.1 - 5 601
U56255.1 - 5 589

View Full Product Details