Gene Transcripts

Gene Symbol
Rqcd1
Entrez Gene ID
58184
Gene Name
rcd1 (required for cell differentiation) homolog 1 (S. pombe)
Gene Aliases
2610007F23Rik, AI593551, Fl10
Chromosome Location
Chr. 1: 74506040 - 74530842 on Build GRCm38
UniGene ID
Mm.291708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021383.5 NP_067358.1 839
GenBank mRNA AF221849.1 - 5 461
AK005025.2 - 5 841
BC050898.1 - 5 640 6401907
BC051948.1 - 678 5708192

View Full Product Details