Gene Transcripts

Gene Symbol
Midn
Entrez Gene ID
59090
Gene Name
midnolin
Gene Aliases
3000003C15Rik
Chromosome Location
Chr. 10: 80148227 - 80158368 on Build GRCm38
UniGene ID
Mm.143813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB036882.1 - 8 1687
AK154535.1 - 7 1628
AK164324.1 - 7 1576
AK164651.1 - 7 1575
BC034719.1 - 8 1657 3710931

View Full Product Details