Gene Transcripts

Gene Symbol
Syt9
Entrez Gene ID
60510
Gene Name
synaptotagmin IX
Gene Aliases
Chromosome Location
Chr. 7: 107370704 - 107548656 on Build GRCm38
UniGene ID
Mm.302793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021889.4 NP_068689.2 7 3327
GenBank mRNA AK089115.1 - 7 3328
AK156631.1 - 7 3328
BC132495.1 - 7 3314 40130772
BC137904.1 - 7 3314 8860923
BC145024.1 - 7 3309

View Full Product Details