Gene Transcripts

Gene Symbol
Naa16
Entrez Gene ID
66897
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases
1300019C06Rik, Narg1l
Chromosome Location
Chr. 14: 79334507 - 79390777 on Build GRCm38
UniGene ID
Mm.24425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025832.2 NP_080108.1 8, 9 968
GenBank mRNA AK005056.1 - 8, 9 978
AK007755.1 - 8, 9 1004
AK028432.1 - 8, 9 1079
AK152498.1 - 8, 9 978
BC052445.1 - 8, 9 968 6412745

View Full Product Details