Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
27418
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
A130067F06Rik, AU015903, BB154892
Chromosome Location
Chr. 6: 31398828 - 31511795 on Build GRCm38
UniGene ID
Mm.238038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013791.2 NP_038819.1 7 781
GenBank mRNA AK037967.1 - 7 782
AK077541.1 - 7 777
AK136123.1 - 4 464
BC013703.1 - 7 765 3668925
U72194.1 - 7 764

View Full Product Details