Gene Transcripts

Gene Symbol
Ube2k
Entrez Gene ID
53323
Gene Name
ubiquitin-conjugating enzyme E2K
Gene Aliases
AW492011, D5Ertd601e, E2-25k, Hip2, Hypg, Lig
Chromosome Location
Chr. 5: 65537238 - 65598989 on Build GRCm38
UniGene ID
Mm.319512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016786.3 NP_058066.2 7 2058
GenBank mRNA AK031055.1 - 5 1758
AK036068.1 - 7 1999
AK048194.1 - 7 2050

View Full Product Details