Gene Transcripts

Gene Symbol
WDR3
Entrez Gene ID
10885
Gene Name
WD repeat domain 3
Gene Aliases
DIP2, UTP12
Chromosome Location
Chr. 1: 117929749 - 117960426 on Build GRCh38
UniGene ID
Hs.310809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006784.2 NP_006775.1 2, 3 232
GenBank mRNA AF083217.1 - 2, 3 213
AK292438.1 - 2, 3 227
BP281064.1 - 2, 3 232

View Full Product Details