Gene Transcripts

Gene Symbol
Tspan5
Entrez Gene ID
56224
Gene Name
tetraspanin 5
Gene Aliases
2810455A09Rik, 4930505M03Rik, NET-4, Tm4sf9
Chromosome Location
Chr. 3: 138742208 - 138904433 on Build GRCm38
UniGene ID
Mm.31927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019571.5 NP_062517.1 4, 5 820
GenBank mRNA AF121344.1 - 4, 5 835
AK013350.1 - 4, 5 785
AK015705.1 - 4, 5 542
AK033543.1 - 4, 5 519
AK187703.1 - 3, 4 334
BC058695.1 - 4, 5 791 6813872

View Full Product Details