Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289774.1 | NP_001276703.1 | 5 | 880 | |
NM_001289775.1 | NP_001276704.1 | 5 | 880 | ||
NM_001289776.1 | NP_001276705.1 | 6 | 1302 | ||
NM_001289777.1 | NP_001276706.1 | 7 | 1127 | ||
NM_027530.3 | NP_081806.1 | 5 | 1248 | ||
GenBank mRNA | AK013185.1 | - | 7 | 1128 | |
AK018184.1 | - | 5 | 1226 | ||
AK083242.1 | - | 5 | 1183 | ||
AK148795.1 | - | 5 | 544 | ||
AK154941.1 | - | 5 | 881 | ||
AK160820.1 | - | 5 | 881 | ||
BC017648.1 | - | 5 | 637 | 4527858 | |
BC049127.1 | - | 5 | 1187 | 6821567 | |
BC058259.1 | - | 6 | 1021 | 6469137 | |
CT010404.1 | - | 5 | 621 |