Gene Transcripts

Gene Symbol
Tor1b
Entrez Gene ID
30934
Gene Name
torsin family 1, member B
Gene Aliases
2610016F05Rik, DQ1, torsinB
Chromosome Location
Chr. 2: 30952938 - 30959015 on Build GRCm38
UniGene ID
Mm.249164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133673.3 NP_598434.2 5 1709
GenBank mRNA AK011416.1 - 1 437
AK028859.1 - 5 1523
AK035602.1 - 1 891
AK054404.1 - 5 1740
AK149754.1 - 5 1729
AK159843.1 - 5 1708
AK171756.1 - 3 2735
BC018456.1 - 5 1708 3582246

View Full Product Details