Gene Transcripts

Gene Symbol
D10Jhu81e
Entrez Gene ID
28295
Gene Name
DNA segment, Chr 10, Johns Hopkins University 81 expressed
Gene Aliases
C21orf33, ES1
Chromosome Location
Chr. 10: 78162067 - 78169768 on Build GRCm38
UniGene ID
Mm.268691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138601.2 NP_613067.1 6, 7 710
GenBank mRNA AK003866.1 - 6, 7 711
AK183350.1 - 1, 2 81
BC013475.1 - 6, 7 703 4237589

View Full Product Details